Drierite Desiccant Bags
  • Desiccant bag will reduce the humidity to a 100°F dew point in 10 hours
  • These bags are dust-tight and will protect for many months
  • Materials packaged with these bags are protected from corrosion
These nonwoven crepe bags are filled with Drierite (anyhydrous calcium sulfate) to completely dry the air within a package or container during shipment or storage. The bags are sturdy and held together by heat seals. Drierite bags also perform well in elevated temperatures and will not release absorbed moisture at less than 350°F.
$123.50 - $320.75USD / Pkg of 750
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Length (in) Width (in)
Availability Pricing
Drierite 60013 Desiccant Bags, 3 g/bag, 750 bags/pk EW-07193-04 Mfr # 60013
COMPARE
3 2 3.75
$195.75USD / Pkg of 750
Drierite 60016 Desiccant Bags, 6 g/bag, 750 bags/pk EW-07193-06 Mfr # 60016
COMPARE
6 2 3.75
$201.00USD / Pkg of 750
Drierite 60011 Desiccant Bags, 1 oz/bag, 200 bags/pk EW-07193-08 Mfr # 60011
COMPARE
1 3.75 4
$123.50USD / Pkg of 200
Drierite 60014 Desiccant Bags, 4 oz/bag, 200 bags/pk EW-07193-12 Mfr # 60014
COMPARE
4 3.75 6.25
$184.00USD / Pkg of 200
Drierite 60018 Desiccant Bags, 8 oz/bag, 100 bags/pk
Always in Stock
EW-07193-14 Mfr # 60018
COMPARE
8 5.75 6.25
$183.00USD / Pkg of 100
Drierite 60012 Desiccant Bags, 2 oz/bag, 400 bags/pk EW-07193-16 Mfr # 60012
COMPARE
2 3.75 4.75
$320.75USD / Pkg of 400