Dynalon Disposable Polystyrene Cuvettes for Visible Spectroscopy
Precision with convenience!
  • % Transmission : maximum from 340 to 800 nm
Use these economical, single-use cuvettes to obtain consistent spectrophotometric data. Available in standard, semi-micro, and fluorometry configurations. Use polystyrene cuvettes for visible spectral analysis.
$235.00 - $285.00USD / Pkg of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
Dynalon Disposable Visible Cuvettes; polystyrene; 4.5 mL; 500/pack EW-06343-10 Mfr # 302195-0002
COMPARE
Polystyrene 4.5 10
$285.00USD / Pkg of 500
Dynalon Square semimicro cuvettes, polystyrene; 1.5 mL capacity, 500/pack EW-06343-20 Mfr # 302205-0002
COMPARE
Polystyrene 1.5 10
$235.00USD / Pkg of 500