Digital Vortex-Genie 2 Shakers
Enhanced vortex action for maximum high-speed touch mixing
  • Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop
Digital Vortex-Genie® 2 Shaker features both touch and hands-free operation with increased timer range. Shake samples from 1 to 99 seconds in touch mode, 1 to 99 minutes in hands-free mode, or continuously in the automatic untimed mode. Digital display provides accuracy, reproducibility, and repeatability. Accepts all standard Genie attachments and accessories and includes pop-off cup and 3" platform.
$667.00 - $674.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Vortex-Genie SI-A236 Digital 2, 500 to 3000 rpm, 120 VAC EW-04724-40 Mfr # SI-A236
COMPARE
120
$674.50USD / Each
Scientific Industries Vortex-Genie SI-A246 Digital 2, 500 to 3000 rpm, 230 VAC EW-04724-45 Mfr # SI-A246
COMPARE
230
$667.00USD / Each