Digital Microplate Genie and Multi-Microplate Genie Shakers
High-speed action for increased mixing efficiency in every well
  • Speed and orbit combine to offer true vortexing action in each well
  • Small vortexing orbit of 1.0 mm for thorough mixing, regardless of sample viscosity
These shakers accept most microplates including 384-well plates, and can be used for PCR plates with optional accessory kit (sold separately). Ideal for use in incubators, cold rooms, or other temperature-sensitive applications. Designed for precise speed and time control. Easily program for on/off pulsing time between 1.1 to 99 minutes, 0 to 99 minutes of mixing, or continuous mode. Multiplate units accept up to four plates or can be expanded to eight with optional accessory tray.
$859.00 - $1,015.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Depth (in) Max Load (lbs) Width (in)
Availability Pricing
Scientific Industries Genie SI-0400A Digital MicroPlate Pulse Shaker, 120V EW-51402-21 Mfr # SI-0400A
COMPARE
6.5 1.1 6.5
$859.00USD / Each
Scientific Industries Genie SI-0401A Digital MicroPlate Pulse Shaker, 230V EW-51402-23 Mfr # SI-0401A
COMPARE
6.5 1.1 6.5
$859.00USD / Each
Scientific Industries Genie SI-4000A Digital Multi-MicroPlate Pulse, 120V EW-51402-25 Mfr # SI-4000A
COMPARE
13.25 1.1 9.4062
$994.00USD / Each
Scientific Industries Genie SI-4001A Digital Multi-MicroPlate Pulse Shaker, 230V EW-51402-28 Mfr # SI-4001A
COMPARE
13.25 1.1 9.4062
$1,015.00USD / Each