Digital Disruptor Genie Shakers
Multidirectional action for simultaneous agitation and vortexing
  • Rapid hands-free disruption of up to 12 microtubes
  • Suitable for use in cold rooms or incubators
Available in analog or digital models, these simple-to-use shakers provide excellent disruption efficiency and are ideal for cell disruption and homogenization. Solid metal casing with rubber feet prevents shaker from "walking" across your benchtop. Included pop-off cup attachment allows use as a convenient vortex mixer for single tubes. Use with lead-free glass Disruptor Beads (sold separately).
$949.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-DD38 Digital Disruptor Shaker, 120 VAC EW-04724-33 Mfr # SI-DD38
COMPARE
120
$949.50USD / Each
Scientific Industries Genie SI-DD48 Digital Disruptor Shaker, 230 VAC, Without Plug. EW-04724-37 Mfr # SI-DD48
COMPARE
230
$949.50USD / Each