Digi-Sense Variable Time Temperature Controllers
Manually set power control dial from 5 to 100% to control the output wattage
This timed control turns the output load on and off and then repeats the cycle. Each position on the knob from LO to HI represents a specific cycle time where the on time ranges from 6 to 100 percent. For example the LO setting represents a total cycle of 60 seconds with an on time of 4 seconds. This type of control can be used to regulate the temperature of resistive loads. The pilot light on the front will glow when the load is on.
$320.00 - $325.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
COMPARE
149 120 Variable- Time Output Controller 1440 Watts, 120 V 50/60 Hz
$320.00USD / Each
COMPARE
149 240 Variable- Time Output Controller, 2400 Watts, 240 V 50/60 Hz
$325.00USD / Each