Digi-Sense Temperature Probe Thermocouple Extension Cables - Miniature
20-gauge (0.032" dia) Thermocouple Wire with PVC/PVC Insulation
  • Maximum temperature: 221° F (105° C).
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
  • Custom designs or changes to this product are available upon request
$39.00 - $89.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Length (ft) Description
Availability Pricing
COMPARE
Mini-Connector 10 Type-T, Extension Cable, Mini Connector, 10ft, 20-Gauge
$39.00USD / Each
COMPARE
Mini-Connector 25 Type-T, Extension Cable, Mini Connector, 25ft, 20-Gauge
$57.50USD / Each
COMPARE
Mini-Connector 10 Type-K, Extension Cable, Mini Connector, 10ft, 20-Gauge
$39.00USD / Each
COMPARE
Mini-Connector 25 Type-K, Extension Cable, Mini Connector, 25ft, 20-Gauge
$55.50USD / Each
COMPARE
Mini-Connector 50 Type-K, Extension Cable, Mini Connector, 50ft, 20-Gauge
$89.00USD / Each
COMPARE
Mini-Connector 10 Type-J, Extension Cable, Mini Connector, 10ft, 20-Gauge
$39.00USD / Each
COMPARE
Mini-Connector 25 Type-J, Extension Cable, Mini Connector, 25ft, 20-Gauge
$57.00USD / Each
COMPARE
Mini-Connector 50 Type-J, Extension Cable, Mini Connector, 50ft, 20-Gauge
$88.00USD / Each
Showing 1- 8 of 8