Digi-Sense Precalibrated Humidity and Temperature Indicators
Designed to gauge proper humidity levels
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Unique environment comfort indicator
  • Displays both temperature and humidly
The Digi-Sense Thermohygrometer Indicators are designed to display temperature, humidity and time. The unique environment comfort indicator lets the user know how the level of humidity is affecting comfort, to high, low or ideal. Advanced features include °F/°C unit selection, min/max reading for temperature and humidity, environment comfort indicator, clock with alarm and date.
Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money
ITEM(S) ON SALE
$46.40USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
COMPARE
1 AAA (included) 90 120
$46.40USD / Each
$58.00USD / Each