Cole-Parmer® Irreversible 5-Point Temperature Labels / Indicators
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.
$27.50 - $84.00USD / Pkg of 25
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (in) Max temperature (° F) Temperature Points (° F)
Availability Pricing
COMPARE
1/2 435 390, 399, 410, 421, 435
$71.50USD / Pkg of 25
COMPARE
1/2 500 450, 466, 480, 489, 500
$71.50USD / Pkg of 25
COMPARE
115 99, 104, 109, 111, 115
$30.70USD / Pkg of 10
COMPARE
160 120, 129, 140, 149, 160
$27.50USD / Pkg of 10
COMPARE
210 171, 180, 180, 199, 210
$27.90USD / Pkg of 10
COMPARE
261 219, 230, 241, 250, 261
$27.90USD / Pkg of 10
COMPARE
309 270, 280, 289, 300, 309
$27.90USD / Pkg of 10
COMPARE
360 320, 331, 340, 351, 360
$27.90USD / Pkg of 10
COMPARE
410 370, 379, 390, 399, 410
$27.90USD / Pkg of 10
COMPARE
480 421, 435, 450, 466, 480
$27.90USD / Pkg of 10
COMPARE
554 480, 489, 500, 536, 554
$31.80USD / Pkg of 10
COMPARE
5/8 130 105, 110, 115, 120, 130
$72.60USD / Pkg of 25
COMPARE
5/8 180 140, 150, 160, 170, 180
$71.50USD / Pkg of 25
COMPARE
5/8 230 190, 200, 210, 220, 230
$80.70USD / Pkg of 25
COMPARE
5/8 280 240, 250, 260, 270, 280
$84.00USD / Pkg of 25
COMPARE
5/8 330 290, 300, 310, 320, 330
$71.50USD / Pkg of 25
COMPARE
5/8 380 340, 350, 360, 370, 380
$71.50USD / Pkg of 25
Showing 1- 17 of 17