Digi-Sense® High-Accuracy Digital Gauges
Durable and reliable digital pressure gauge
  • High Accuracy with high repeatability
  • Backlilght and Zero-Reset Standard
  • Optional line powered with 4/20mA available & NPN limit switch
These digital pressure and vacuum gauges are an ideal alternative to your mechanical gauges! These gauges offer high accuracy plus the convenience of a digital display. The semiconductor sensing element measures pressure with ±0.25% or ±0.5% accuracy (depending on the range), and lasts for millions of cycles. These gauges come standard with Zero-Reset, Backlight, and Auto-Shutoff to help prolong battery life. Peak hold is standard on all pressure units.
As an option, some models can also come with a 4/20mA transmitter and NPN limit switch. These units are line powered with 8-30VDC.
ITEM(S) ON SALE
$280.50 - $344.25USD / Each
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±0.50% full-scale 30 0.01 psi
$294.95USD / Each
$347.00USD / Each
COMPARE
±0.50% full-scale 50 0.01 psi
$289.00USD / Each
$340.00USD / Each
COMPARE
±0.25% full-scale 100 0.1 psi
$283.05USD / Each
$333.00USD / Each
COMPARE
±0.25% full-scale 200 0.1 psi
$280.50USD / Each
$330.00USD / Each
COMPARE
±0.25% full-scale 300 0.1 psi
$289.00USD / Each
$340.00USD / Each
COMPARE
±0.25% full-scale 500 0.1 psi
$289.00USD / Each
$340.00USD / Each
COMPARE
±0.25% full-scale 1000 1 psi
$284.75USD / Each
$335.00USD / Each
COMPARE
±0.25% full-scale 3000 1 psi
$289.00USD / Each
$340.00USD / Each
COMPARE
±0.25% full-scale 5000 1 psi
$289.00USD / Each
$340.00USD / Each
COMPARE
±0.25% full-scale 2000 1 psig
$289.00USD / Each
$340.00USD / Each
COMPARE
±0.5% full-scale 15 0.01 psig
$281.35USD / Each
$331.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 0.01 psi
$306.00USD / Each
$360.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 0.01\" Hg
$306.00USD / Each
$360.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 15 0.01 psi
$313.22USD / Each
$368.50USD / Each
COMPARE
±0.5% full-scale ±1 digit 100 0.1 psi
$297.50USD / Each
$350.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 200 0.1 psi
$310.25USD / Each
$365.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 5 0.01 psi
$289.00USD / Each
$340.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 5 0.01 psi
$344.25USD / Each
$405.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 15 0.01 psi
$344.25USD / Each
$405.00USD / Each
COMPARE
±0.5% full-scale ±1 digit 100 0.01 psi
$344.25USD / Each
$405.00USD / Each
Showing 1 - 20 of 23