Digi-Sense Dual Scale 2" Dial Stainless Steel Pocket Thermometers
Choose the dial size and temperature range to meet your specific needs
  • Factory tested to three points with a ±1% accuracy
  • Minimum immersion: liquids 2" (50 mm), gases 4" (100 mm)
  • Hermetically sealed dial stops internal fogging
  • Recalibrate using external adjustment nut
  • 304 Stainless Steel dial case and stem
  • Shock resistant
$29.20USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Dial Size (in) Stem Length (in) Description
Availability Pricing
COMPARE
2 8 S.S. Bimetalal Pkt Thermo 25-125F (-4-52C) 2\" Dial, Poly Lens, 8\" Stem, 1F Div
$29.20USD / Each
COMPARE
2 8 S.S. Bimetalal Pkt Thermo 0-220F 2\" Dial, Poly Lens, 8\" Stem, 2F Div
$29.20USD / Each
COMPARE
2 8 S.S. Bimetalal Pkt Thermo 50-550F 2\" Dial, Poly Lens, 8\" Stem, 5F Div
$29.20USD / Each