Digi-Sense® Digital Industrial Pressure Gauges
Signal conditioned for temperature
  • ±1% full-scale accuracy
  • Read pressure in psi, Bar, or MPA
  • Laser-welded stainless steel design
The all laser-welded 316L stainless steel gauge is designed for superior media isolation and can be used in a wide variety of applications including marine, refrigeration, pumps, hydraulics, pneumatics, and wastewater.
Simple keypad operation features auto zero and peak hold, and can set display to show pressure in psi, bar, and MPa. Program features also include a display power down after three minutes to save battery life.
Models 68950-58, -59, and -60 are designed for pneumatic use only; they feature a 304L stainless steel pressure port. These gauges should only be used with dry air and noncorrosive gases.
ITEM(S) ON SALE
$130.47 - $206.55USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Resolution Description
Availability Pricing
COMPARE
5 0.01 psi Digital Pressure Gauge, 0 to 5 psi, 2.5\" dia, 1/4\" NPT(M)
$192.95USD / Each
$227.00USD / Each
COMPARE
30 0.01 psi Digital Pressure Gauge, 0 to 30 psi, 2.5\" dia, 1/4\" NPT(M)
$197.62USD / Each
$232.50USD / Each
COMPARE
100 0.1 psi Digital Pressure Gauge, 0 to 100 psi, 2.5\" dia, 1/4\" NPT(M)
$192.95USD / Each
$227.00USD / Each
COMPARE
200 0.1 psi Digital Pressure Gauge, 0 to 200 psi, 2.5\" dia, 1/4\" NPT(M)
$190.40USD / Each
$224.00USD / Each
COMPARE
500 0.1 psi Digital Pressure Gauge, 0 to 500 psi, 2.5\" dia, 1/4\" NPT(M)
$192.95USD / Each
$227.00USD / Each
COMPARE
1000 1 psi Digital Pressure Gauge, 0 to 1000 psi, 2.5\" dia, 1/4\" NPT(M)
$190.40USD / Each
$224.00USD / Each
COMPARE
3000 1 psi Digital Pressure Gauge, 0 to 3000 psi, 2.5\" dia, 1/4\" NPT(M)
$197.62USD / Each
$232.50USD / Each
COMPARE
5000 1 psi Digital Pressure Gauge, 0 to 5000 psi, 2.5\" dia, 1/4\" NPT(M)
$206.55USD / Each
$243.00USD / Each
COMPARE
30 0.1 psi Digital Pressure Gauge, 0 to 30 psi, 2.5\" dia, 1/4\" NPT(M), air/gas only
$134.72USD / Each
$158.50USD / Each
COMPARE
100 0.1 psi Digital Pressure Gauge, 0 to 100 psi, 2.5\" dia, 1/4\" NPT(M), air/gas only
$130.47USD / Each
$153.50USD / Each
COMPARE
300 0.1 psi Digital Pressure Gauge, 0 to 300 psi, 2.5\" dia, 1/4\" NPT(M), air/gas only
$132.17USD / Each
$155.50USD / Each
Showing 1- 11 of 11