Digi-Sense® Advanced Precalibrated Thermocouple Thermometers
Perfect for the lab to production floor
  • Reliable measurements, guaranteed!
  • Choose from meter or meter system—both include calibration
  • Heavy duty construction to protect against knocks and jars.
  • Super large LCD for easy viewing of data
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
The Digi-Sense Professional Thermocouple Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours.
Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you hundreds of dollars and extra waiting time to calibrate separately.
ITEM(S) ON SALE
$91.20 - $184.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Logging Number Of Probe Channels Description
Availability Pricing
COMPARE
No 1 Single-Input Thermocouple Thermometer with NIST-Traceable Calibration
$91.20USD / Each
$114.00USD / Each
COMPARE
Yes 2 Dual-Input Data Logging Thermocouple Thermometer with NIST-Traceable Calibration
$167.20USD / Each
$209.00USD / Each
COMPARE
No 1 Single-Input Thermocouple Thermometer System with NIST-Traceable Calibration
$184.00USD / Pkg of 1
$230.00USD / Pkg of 1