Digi-Sense 1" Dial Stainless Steel Pocket Thermometers
Choose the dial size and temperature range to meet your specific needs
  • Factory tested to three points with a ±1% accuracy
  • Minimum immersion: liquids 2" (50 mm), gases 4" (100 mm)
  • Hermetically sealed dial stops internal fogging
  • Recalibrate using external adjustment nut
  • 304 Stainless Steel dial case and stem
  • Shock resistant
$12.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Dial Size (in) Stem Length (in) Description
Availability Pricing
COMPARE
1 5 S.S. Bimetalal Pocket Thermometer 0-250C, 1\" Dial, Poly Lens, 5\" Stem, 1C Div
$12.50USD / Each
COMPARE
1 5 S.S. Bimetalal Pocket Thermometer 50-550F, 1\" Dial, Poly Lens, 5\" Stem, 5F Div
$12.50USD / Each