Diba Omnifit® Reservoir Filters and Bubblers
Push-fit onto a 1/8" OD tube for a quick connection
  • Optimize solvent filtration with filters and bubblers
These reservoir filters and bubbler provide simple and effective solvent filtration. A 10 µm filter ensures even the smallest particulates are removed from the solvent supply line. Bottom-of-bottle filters have slots in the body allowing virtually all liquid to be removed from the bottle, minimizing waste. Select from PTFE body/membrane or PTFE body/PE membrane filters. Filter bubbler, suitable for filtration or sparging applications, comes with a 316 SS filter.
$58.80 - $63.80USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Membrane Material
Availability Pricing
Diba Omnifit® Bottom-of-Bottle Filter for GL32 /38-430 Bottle, 10 µm PTFE; 1/ea EW-21943-32 Mfr # 009F-32
COMPARE
For GL32/38-430 bottle PTFE
$58.80USD / Each
Diba Omnifit® Bottom-of-Bottle Filter for GL45 Bottle, 10 µm PTFE; 1/ea EW-21943-34 Mfr # 009F-45
COMPARE
For GL45 bottle PTFE
$63.50USD / Each
Diba Omnifit® Filter Bubbler, 316 Stainless Steel; 1/ea EW-21943-42 Mfr # 009SFB
COMPARE
For any bottle 316 Stainless steel
$63.80USD / Each