Diba Omnifit® High-Pressure Rotary Valves

Rated for use up to 500 psi (33 bar)
  • Choose from 4-or 5-port valves
Use the high-pressure, chemically inert manual rotary valves with a variety of flow configurations for sampling, flow splitting, mixing, and fluid-line interconnection. Valves are constructed from PTFE with a PCTFE rotor. They accept 1/4"-28 UNF(M) fittings and feature a 0.8 mm bore and M3 mounting holes. Select from 4- or 5-port versions. Use plug 21940-04 to seal any unused ports.
$330.00 - $355.25USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Pattern Inlet Port
Availability Pricing
Diba Omnifit® High-Pressure Valve with 1/4"-28 UNF(F), 4-port, 4-way, 90° flow; 1/ea EW-21939-24 Mfr # 001120
COMPARE
Connect two ports at 90° to each other 4 ports
$330.00USD / Each
Diba Omnifit® High-Pressure Valve with 1/4"-28 UNF(F), 4-port, 3-way, tee flow; 1/ea EW-21939-25 Mfr # 001121
COMPARE
Connect three ports in a \"T\" 4 ports
$330.00USD / Each
Diba Omnifit® High-Pressure Valve with 1/4"-28 UNF(F), 4-port, 2-way, adjacent flow; 1/ea EW-21939-26 Mfr # 001122
COMPARE
Connect adjacent pairs of ports at the same time 4 ports
$355.25USD / Each
Diba Omnifit® High-Pressure Valve with 1/4"-28 UNF(F), 5-port, 5-way, 1 common port; 1/ea EW-21939-27 Mfr # 001126
COMPARE
Connect one common port to any one of four other ports 5 ports
$333.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat