Diba Omnifit® Bubble Traps
Remove bubbles with or without vacuum assistance
  • Constructed with durable PTFE membrane
Effectively remove bubbles* from aqueous solutions inline or downstream with bubble traps. Traps feature ¼"-28 UNF(F) port and come with 10 µm PTFE filter. The high-flow trap features a vacuum port on the atmosphere (dry) side of the membrane. When vacuum is applied, the pressure differential between the system and atmosphere side increases (must not exceed 30 psi/2.1 bar), causing bubbles to be removed from the liquid and permitting increased liquid flow.
$118.75 - $202.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate Material
Availability Pricing
Diba Omnifit® Standard Flow Bubble Trap, PEEK™, 10 µm PTFE filter; 1/ea EW-21940-38 Mfr # 006BT
COMPARE
6 mL/min (max) PEEK™ body, PTFE membrane
$202.00USD / Each
Diba Omnifit® High Flow Bubble Trap, PVC, 10 µm PTFE filter; 1/ea
Always in Stock
EW-21940-39 Mfr # 006BT-HF
COMPARE
60 mL/min (max) PVC body, PTFE membrane
$118.75USD / Each