Delmhorst TechCheck Plus Moisture Meters
Simple and versatile – 2-in-1 moisture meter
  • Two measuring modes: built-in pins, or nondestructive pinless scanner
  • Expand capabilities with optional electrodes
  • Drywall scale display
  • User-adjustable alarm lets you know when a preselected moisture threshold is reached
TechCheck Plus is the simplest and most versatile 2-in-1 moisture meter designed with the technician in mind. Meter offers advanced features such as a large backlit display, user-adjustable alarm, data Hold, drywall scale display, and multiple languages that make even the most demanding inspection simple.
This easy-to-use meter features seamless integration of pin and pinless moisture measurement modes and a streamlined user interface. The pin mode is used to locate the exact source of moisture. The contact pins mounted on top of the meter provide 5/16" (0,8cm) penetration for testing drywall and wood materials The scan mode provides a numerical reference scale that allows you to quickly scan large areas and identify elevated moisture levels
$664.00 - $717.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Measurement Type
Availability Pricing
Delmhorst TECHCHECK Moisture Meter, Basic Package
Always in Stock
EW-59820-61 Mfr # TECHCHECKPLUSW/CS
COMPARE
1 9V (included) Combination
$664.00USD / Each
Delmhorst TECHCHECK/PKG Techcheck Plus Moisture Meter Insulation Package EW-59820-63 Mfr # TECHCHECKPLUS/PKG
COMPARE
1 9V (included) Combination
$717.00USD / Each