Delmhorst J-4 Analog Wood Moisture Meter
Ideal for testing moisture content in wood
  • Easy-to-read analog display
  • Built-in calibration check verifies accuracy of readings
  • Rugged construction ensures years of reliable use
The user-friendly analog wood moisture meter accurately measures moisture content in wood over a range of 6 to 30%. Contact pins mounted on top of the meter offer 5/16” penetration. A built-in connector allows an electrode to be attached to the meter for checking wood over an inch thick.
$391.00 - $570.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Delmhorst J-4W/CS Analog Wood Moisture Meter EW-59823-29 Mfr # J-4W/CS
COMPARE
Analog Wood Moisture Meter
$391.00USD / Each
Delmhorst J-4/PKG Analog Wood Moisture Meter Kit EW-59823-30 Mfr # J-4/PKG
COMPARE
Analog Wood Moisture Meter Kit
$570.00USD / Each