Delmhorst F-2000 Digital Hay Moisture Meter
Safely store hay bales
  • Ensure correct storage conditions to inhibit fungal rot and avert spontaneous combustion
  • Store up to 100 readings with average and maximum
  • Audible alarm warns you when moisture content exceeds the set point
This moisture meter will help ensure hay is baled and stored at the optimum moisture content to avoid spoilage and prevent the dangerous temperature increase associated with rot which can turn your crop into a fire hazard.
A variety of electrodes are available to customize the meter to your application including three lengths of bale prods, a short pin prod for testing hay in the windrow, and a bale sensor for continuously monitoring the bale chamber from the cab.
$359.00 - $552.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Delmhorst F-2000 F-2000 Digital Hay Moisture Meter Only EW-59823-06 Mfr # F-2000
COMPARE
F-2000 Digital Hay Moisture Meter Only
$359.00USD / Each
Delmhorst F-2000/1235 F-2000/1235 Digital Hay Moisture Meter with 10" Probe EW-59823-07 Mfr # F-2000/1235
COMPARE
F-2000/1235 Digital Hay Moisture Meter with 10\" Probe
$418.00USD / Each
Delmhorst F-2000/PKG F-2000/PKG Digital Hay Moisture Meter Kit EW-59823-08 Mfr # F-2000/PKG
COMPARE
F-2000/PKG Digital Hay Moisture Meter Kit
$552.00USD / Each