Cytoseal Mounting Media
  • Not for sale outside of the United States
Clear, colorless media contains an anti-oxidant to prevent yellowing or fading of stained sections. Media come in an easy-to-use dispenser bottle — prevent spills while promoting precise dispensing. Feature the same refractive index as immersion oil. Sold in case of 12.
$234.00 - $296.75USD / Case of 12
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Description
Availability Pricing
Richard-Allan Scientific Cytoseal Cytoseal 60 Histology Mounting Media, 4 oz Bottles; 12/Cs EW-48802-01 Mfr # 8310-4
COMPARE
118 Cytoseal 60 Histology Mounting Media, 4 oz Bottles; 12/Cs
$296.75USD / Case of 12
Richard-Allan Scientific Cytoseal Cytoseal 280 Histology Mounting Media, 4 oz Bottles; 12/Cs EW-48802-02 Mfr # 8311-4
COMPARE
118 Cytoseal 280 Histology Mounting Media, 4 oz Bottles; 12/Cs
$234.00USD / Case of 12