CYTIVA Whatman™ Disposable Capsule Filters, PTFE Membrane
PTFE membrane is ideal for venting, in-line filtration, and isolation (gas pass, liquids, and aerosol stopped)
  • Autoclavable
Capsules feature sturdy PP housings with fused seals—no glues or adhesives to contaminate filtrate. Capsules include a luer lock vent to guarantee that no filtration area is wasted. Use vent to draw solution into the capsule or let air escape from the capsule or sample port.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$139.00 - $196.25USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Whatman 6700-3602 Polycap TF PTFE Capsule Filter, 0.2um, 500cm2, 1/4-3/8" ID; 1/Pk EW-29700-00 Mfr # 6700-3602
COMPARE
Polycap TF PTFE Capsule Filter, 0.2um, 500cm2, 1/4-3/8\" ID; 1/Pk
$139.00USD / Each
Whatman 6700-3610 Polycap TF PTFE Capsule Filter, 1.0um, 500cm2, 1/4-3/8" ID; 1/Pk EW-29700-02 Mfr # 6700-3610
COMPARE
Polycap TF PTFE Capsule Filter, 1.0um, 500cm2, 1/4-3/8\" ID; 1/Pk
$169.50USD / Each
Whatman 6700-7501 Polycap TF PTFE Capsule Filter, 0.1um, 1000cm2, 1/4-3/8" ID; 1/Pk EW-29700-10 Mfr # 6700-7501
COMPARE
Polycap TF PTFE Capsule Filter, 0.1um, 1000cm2, 1/4-3/8\" ID; 1/Pk
$196.25USD / Each
Whatman 6700-7502 Polycap TF PTFE Capsule Filter, 0.2um, 1000cm2, 1/4-3/8" ID; 1/Pk EW-29700-12 Mfr # 6700-7502
COMPARE
Polycap TF PTFE Capsule Filter, 0.2um, 1000cm2, 1/4-3/8\" ID; 1/Pk
$181.00USD / Each
Whatman 6700-7504 Polycap TF PTFE Capsule Filter, 0.45um, 1000cm2, 1/4-3/8" ID; 1/Pk EW-29700-14 Mfr # 6700-7504
COMPARE
Polycap TF PTFE Capsule Filter, 0.45um, 1000cm2, 1/4-3/8\" ID; 1/Pk
$193.75USD / Each
Whatman 6701-7510 Polycap TF PTFE Capsule Filter, 1.0um, 1000cm2, 1/2" ID; 1/Pk EW-29700-16 Mfr # 6701-7510
COMPARE
Polycap TF PTFE Capsule Filter, 1.0um, 1000cm2, 1/2\" ID; 1/Pk
$172.25USD / Each