CR Magnetics Current Ground Fault and DC Sensing Relays
These UL listed Current Sensing Relays are designed to monitor current levels in AC conductors, and provide a dry relay contact activation when the current rises above or falls below a user adjusted set point. The CR4395 series is for general applications, and the CR7310 series is for ground fault measurement and the CR5395 series, Direct Current Sensing relay provides a precision and cost effective method for monitoring DC current.
FEATURES
  • Variable trip point and time delay
  • Monitors currents from 10mA to 60 AC Amps (CR7310 and CR4395)
  • Monitors currents from 1.0 Adc to 100 Adc (CR5395)
  • Bi-polar (CR5395)
  • Electrical isolation between circuits
  • Output relay rated up to 20 Amps
  • LED relay status indicator
  • Dead band prevents relay chatter
  • Calibrated dial
  • External current transformers available
$248.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Load Rating Max Current (amps) Relay Type
Availability Pricing
CR Magnetics CR7310 Ground Fault Sensing Relay 120Vac Trip Range 0.1 to 1.0 A EW-68480-37 Mfr # CR7310-EH-120-.11-X-CD-ELR-I
COMPARE
0.1 to 1.0 A 20
$248.50USD / Each