CP Wet/Wet Differential Multiconfigurable Pressure Transducers
Measure eight pressure ranges in a single unit
  • Choose models with LCD for on-site indication of pressure
These feature-rich, multiconfigurable wet-to-wet differential pressure transducers provides you with field-accessible pressure ranging, choice of output, and zeroing. Each unit has four unidirectional and four bidirectional switch-selectable ranges — read positive and negative gauge and differential pressure ranges, giving you eight pressure ranges in one transducer! All controls are contained in a NEMA 4 rated, cast aluminum housing that is easily accessible in the field.
ITEM(S) ON SALE
$817.70USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±1% full-scale; ±2% full-scale at lowest psi rating
$817.70USD / Each
$962.00USD / Each