Costar® Sterile Multiwell Tissue Culture Plates with TC Surface Treatment
Optically clear, virgin polystyrene is treated for optimal cell attachment
  • Stacking beads and uniform footprints facilitate stacking and handling
  • Individual alphanumeric codes for well identification
  • Lids feature individual condensation rings to reduce well-to-well contamination
  • Gamma radiation sterilized and certified pyrogen-free
These multiwell plates feature tissue culture (TC) surface treatment for optimal cell attachment. Serrations on the plate skirt enables you to easily distinguish plates from lids when handling stacks of clusters.
$230.00 - $675.00USD / Case of 100
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Packaging Well Volume (mL)
Availability Pricing
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 6-Well, 9.5 cm2; 100/Cs EW-01959-00 Mfr # 3506
COMPARE
6 5 Plates/Pack, 100 Plates/Case 16.8
$450.00USD / Case of 100
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 6-Well, 9.5 cm2; 50/Cs EW-01959-02 Mfr # 3516
COMPARE
6 1 Plate/Pack, 50 Plates/Case 16.8
$230.00USD / Case of 50
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 12-Well, 3.8 cm2; 100/Cs EW-01959-04 Mfr # 3512
COMPARE
12 5 Plates/Pack, 100 Plates/Case 6.9
$520.00USD / Case of 100
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 12-Well, 3.8 cm2; 50/Cs EW-01959-06 Mfr # 3513
COMPARE
12 1 Plate/Pack, 50 Plates/Case 6.9
$275.00USD / Case of 50
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 24-Well, 1.9 cm2; 100/Cs EW-01959-08 Mfr # 3524
COMPARE
24 1 Plate/Pack, 100 Plates/Case 3.4
$575.00USD / Case of 100
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 24-Well, 1.9 cm2; 50/Cs EW-01959-10 Mfr # 3526
COMPARE
24 1 Plate/Pack, 50 Plates/Case 3.4
$290.00USD / Case of 50
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 48-Well, 0.95 cm2; 100/Cs EW-01959-12 Mfr # 3548
COMPARE
48 1 Plate/Pack, 100 Plates/Case 1.6
$675.00USD / Case of 100
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 96-Well, Flat Bottom; 5/Pk, 50/Cs EW-01959-20 Mfr # 3585
COMPARE
96 5 Plates/Pack, 50 Plates/Case 0.36
$380.00USD / Case of 50
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 96-Well, Flat Bottom; 1/Pk, 50/Cs EW-01959-22 Mfr # 3596
COMPARE
96 1 Plate/Pack, 50 Plates/Case 0.36
$395.00USD / Case of 50
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 96-Well, Flat Bottom; 10/Pk, 50/Cs EW-01959-24 Mfr # 3997
COMPARE
96 10 Plates/Pack, 50 Plates/Case 0.36
$355.00USD / Case of 50
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 96-Well, Flat Bottom; 5/Pk, 100/Cs EW-01959-26 Mfr # 3598
COMPARE
96 5 Plates/Pack, 100 Plates/Case 0.36
$605.00USD / Case of 100
Costar Cell Culture Multiwell Plate with Lid, Sterile, TC-Treated, 96-Well, Round Bottom; 1/Pk, 50/Cs EW-01959-28 Mfr # 3799
COMPARE
96 1 Plate/Pack, 50 Plates/Case 0.33
$425.00USD / Case of 50
Showing 1- 12 of 12