Costar® 96-Well Microplates, Polypropylene
Uniform skirt heights for greater robotic gripping surface
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance
  • Certified RNase and DNase free
Standard microplates are built to industry standard footprint and compatible with automated systems. Deep-well plates are ideal for higher-volume storage and assay applications. Lids are not included, for standard microplates and 1.0-mL deep well plates, use storage mat 01729-22 and applicator 01729-24 sold separately.
$250.00 - $1,120.00USD / Case of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Plate Type Sterile Well Shape
Availability Pricing
Costar 96-Well Microplate, Standard Well, Non-Sterile, V-Bottom, 0.32 mL; 100/Cs EW-01728-80 Mfr # 3363
COMPARE
Standard Well No V-Bottom
$425.00USD / Case of 100
Costar 96-Well Microplate, Standard Well, Non-Sterile, Flat Well, 0.36 mL; 100/Cs EW-01728-81 Mfr # 3364
COMPARE
Standard Well No Flat
$425.00USD / Case of 100
Costar 96-Well Microplate, Standard Well, Non-Sterile, Round Well, 0.36 mL; 100/Cs EW-01728-82 Mfr # 3365
COMPARE
Standard Well No Round
$420.00USD / Case of 100
Costar 96-Well Microplate, Deep Well, Sterile, Round Bottom, 1 mL; 25/Cs EW-01729-02 Mfr # 3958
COMPARE
Deep Well Yes Round
$250.00USD / Case of 25
Costar 96-Well Microplate, Deep Well, Sterile, V-Bottom, 2 mL; 25/Cs EW-01729-03 Mfr # 3960
COMPARE
Deep Well Yes V-Bottom
$315.00USD / Case of 25
Costar 96-Well Microplate, Deep Well, Non-Sterile, Round Bottom, 1 mL; 100/Cs EW-01729-04 Mfr # 3959
COMPARE
Deep Well No Round
$890.00USD / Case of 100
Costar 96-Well Microplate, Deep Well, Non-Sterile, V-Bottom, 2 mL; 100/Cs EW-01729-05 Mfr # 3961
COMPARE
Deep Well No V-Bottom
$1,120.00USD / Case of 100
Showing 1- 7 of 7