Corning® Rectangular EVA-Foam Ice Pans
Recyclable, inert, and nontoxic
  • Highly insulative, unbreakable, lightweight and leakproof
  • Textured finish on bottom prevents slipping on your benchtop
  • Stackable design saves space
  • Available in a range of bright colors with or without lids
Made of ethylene-vinyl acetate (EVA) foam, these highly insulative containers will not sweat or leak. They are ideal for use with ice, dry ice (-78°C), liquid nitrogen (-196°C), alcohol or saline solutions, or warm solutions up to 93°C.
$90.00 - $225.00USD / Each
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Color
Availability Pricing
Corning EVA-Foam Ice Pan, Green, 9 L; 1/Each EW-04392-93 Mfr # 432093
COMPARE
9 Green
$195.00USD / Each
Corning EVA-Foam Ice Pan, Blue, 9 L; 1/Each EW-04392-94 Mfr # 432094
COMPARE
9 Blue
$195.00USD / Each
Corning EVA-Foam Ice Pan, Red, 9 L; 1/Each EW-04392-95 Mfr # 432095
COMPARE
9 Red
$195.00USD / Each
Corning EVA-Foam Ice Pan, Orange, 9 L; 1/Each EW-04392-96 Mfr # 432096
COMPARE
9 Orange
$195.00USD / Each
Corning EVA-Foam Ice Pan, Lime, 9 L; 1/Each EW-04392-97 Mfr # 432097
COMPARE
9 Lime
$195.00USD / Each
Corning EVA-Foam Ice Pan, Pink, 9 L; 1/Each EW-04392-98 Mfr # 432098
COMPARE
9 Pink
$195.00USD / Each
Corning EVA-Foam Ice Pan, Purple, 9 L; 1/Each EW-04392-99 Mfr # 432099
COMPARE
9 Purple
$195.00USD / Each
Corning EVA-Foam Ice Pan with Lid, Blue, 9 L; 1/Each EW-04393-00 Mfr # 432100
COMPARE
9 Blue with Lid
$225.00USD / Each
Corning EVA-Foam Ice Pan with Lid, Green, 9 L; 1/Each EW-04393-01 Mfr # 432101
COMPARE
9 Green with Lid
$225.00USD / Each
Corning EVA-Foam Ice Pan with Lid, Purple, 9 L; 1/Each EW-04393-02 Mfr # 432102
COMPARE
9 Purple with Lid
$225.00USD / Each
Corning EVA-Foam Ice Pan, Green, 4 L; 1/Each EW-04393-03 Mfr # 432103
COMPARE
4 Green
$145.00USD / Each
Corning EVA-Foam Ice Pan, Blue, 4 L; 1/Each EW-04393-04 Mfr # 432104
COMPARE
4 Blue
$145.00USD / Each
Corning EVA-Foam Ice Pan, Red, 4 L; 1/Each EW-04393-05 Mfr # 432105
COMPARE
4 Red
$145.00USD / Each
Corning EVA-Foam Ice Pan, Orange, 4 L; 1/Each EW-04393-06 Mfr # 432106
COMPARE
4 Orange
$145.00USD / Each
Corning EVA-Foam Ice Pan, Lime, 4 L; 1/Each EW-04393-07 Mfr # 432107
COMPARE
1 Lime
$145.00USD / Each
Corning EVA-Foam Ice Pan, Pink, 4 L; 1/Each EW-04393-08 Mfr # 432108
COMPARE
4 Pink
$145.00USD / Each
Corning EVA-Foam Ice Pan, Purple, 4 L; 1/Each EW-04393-09 Mfr # 432109
COMPARE
4 Purple
$145.00USD / Each
Corning EVA-Foam Ice Pan with Lid, Blue, 4 L; 1/Each EW-04393-10 Mfr # 432110
COMPARE
4 Blue with Lid
$165.00USD / Each
Corning EVA-Foam Ice Pan with Lid, Orange, 4 L; 1/Each EW-04393-11 Mfr # 432111
COMPARE
4 Orange with Lid
$165.00USD / Each
Corning EVA-Foam Ice Pan with Lid, Green, 4 L; 1/Each EW-04393-12 Mfr # 432112
COMPARE
4 Green with Lid
$165.00USD / Each
Showing 1 - 20 of 29