Corning Disposable Bottletop Filters
Ideal for filtering cell culture media, biological fluids, and aqueous solutions
  • Large membrane diameters (50, 70, and 90mm) and glass prefilters increase flow rates
  • Provided sterile and certified nonpyrogenic
Polystyrene funnel with polyethylene neck adapter is designed for use with wide-mouth (45mm) polystyrene or glass storage bottles. All bottle-top filters are color-coded and printed with the membrane type and pore size for easy identification.
$126.25 - $358.75USD / Case of 12
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Corning 430514 Disposable Bottletop Filter; 500 mL 0.45µm EW-01937-14 Mfr # 430514
COMPARE
Disposable Bottletop Filter; 500 mL 0.45µm
$126.25USD / Case of 12
Corning 431161 Disposable Bottletop Filter; 150 mL volume; PES, membrane; 48/pack EW-29530-32 Mfr # 431161
COMPARE
Disposable Bottletop Filter; 150 mL volume; PES, membrane; 48/pack
$343.50USD / Case of 48
Corning 430626 Disposable Bottletop Filter; 150 mL volume; CA, membrane; 48/pack EW-29530-34 Mfr # 430626
COMPARE
Disposable Bottletop Filter; 150 mL volume; CA, membrane; 48/pack
$358.75USD / Case of 48
Corning 431118 Disposable Bottletop Filter; 500 mL volume; PES, membrane; 12/pack EW-29530-42 Mfr # 431118
COMPARE
Disposable Bottletop Filter; 500 mL volume; PES, membrane; 12/pack
$126.25USD / Case of 12
Corning 430513 Disposable Bottletop Filter; 500 mL volume; CA, membrane; 12/pack EW-29530-44 Mfr # 430513
COMPARE
Disposable Bottletop Filter; 500 mL volume; CA, membrane; 12/pack
$126.25USD / Case of 12
Corning 430758 Disposable Bottletop Filter; 500 mL volume; CN, membrane; 12/pack EW-29530-47 Mfr # 430758
COMPARE
Disposable Bottletop Filter; 500 mL volume; CN, membrane; 12/pack
$202.00USD / Pkg of 12
Corning 430049 Disposable Bottletop Filter; 500 mL volume; NYL, membrane; 12/pack EW-29530-48 Mfr # 430049
COMPARE
Disposable Bottletop Filter; 500 mL volume; NYL, membrane; 12/pack
$136.50USD / Case of 12
Corning 431174 Disposable Bottletop Filter; 1000 mL volume; PES, membrane; 12/pack EW-29530-52 Mfr # 431174
COMPARE
Disposable Bottletop Filter; 1000 mL volume; PES, membrane; 12/pack
$202.00USD / Case of 12
Corning 430015 Disposable Bottletop Filter; 1000 mL volume; CA, membrane; 12/pack EW-29530-54 Mfr # 430015
COMPARE
Disposable Bottletop Filter; 1000 mL volume; CA, membrane; 12/pack
$212.25USD / Case of 12
Showing 1- 9 of 9