Corning® CoolCell® Containers
Alcohol-free cell freezing reduces messes and costs
  • Solid thermoconductive core eliminates the hassle of IPA changes
  • Control the rate of freezing to -1°C/minute in a -80°C freezer to maintain cell viability
  • Highly reproducible cooling profile ensures repeatable results
This freezing system constructed of uniform-density cross-linked polyethylene foam with a solid-state core cools at a consistent rate, providing living samples the best opportunity to remain viable throughout the freezing and storage process. Easy to use—load your sample tubes and place in the freezer. CoolCell containers rapidly return to room temperature when removed from the freezer, making them easy to handle.
Use filler vials (order separately) when you need to freeze less than a full load of samples to maintain a consistent cooling profile. Order additional vial modules to quickly change out all 30 vials at once. Modules are compatible with standard 5" x 5" x 2" (12.7 x 12.7 x 5.1 cm) cryogenic storage boxes.
$185.00 - $630.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tubes Held Tube Size
Availability Pricing
Corning CoolCell Freezing System for Cryogenic Vials, purple, 12 unexposed vials; 1/ea EW-04392-00 Mfr # 432000
COMPARE
Purple 12 1.0 to 2.0 mL, cryogenic
$185.00USD / Each
Corning LX CoolCell Freezing System for Cryogenic Vials, purple, 12 exposed vials; 1/ea EW-04392-01 Mfr # 432001
COMPARE
Purple 12 1.0 to 2.0 mL, cryogenic
$230.00USD / Each
Corning LX CoolCell Freezing System for Cryogenic Vials, green, 12 exposed vials; 1/ea EW-04392-02 Mfr # 432002
COMPARE
Green 12 1.0 to 2.0 mL, cryogenic
$230.00USD / Each
Corning LX CoolCell Freezing System for Cryogenic Vials, orange, 12 exposed vials; 1/ea EW-04392-03 Mfr # 432003
COMPARE
Orange 12 1.0 to 2.0 mL, cryogenic
$230.00USD / Each
Corning LX CoolCell Freezing System for Cryogenic Vials, pink, 12 exposed vials; 1/ea EW-04392-04 Mfr # 432004
COMPARE
Pink 12 1.0 to 2.0 mL, cryogenic
$230.00USD / Each
Corning LX CoolCell Freezing System f/ Cryogenic Vials, purple, 5mL, 12 exposed vials; 1/ea EW-04392-05 Mfr # 432005
COMPARE
Purple 12 5.0 mL, cryogenic
$310.00USD / Each
Corning FTS30 CoolCell Freezing System for Cryogenic Vials, purple, 30 exposed vials; 1/ea EW-04392-06 Mfr # 432006
COMPARE
Purple 30 1.0 to 2.0 mL, cryogenic
$630.00USD / Each
Corning FTS30 CoolCell Freezing System for Cryogenic Vials, orange, 30 exposed vials; 1/ea EW-04392-07 Mfr # 432007
COMPARE
Orange 30 1.0 to 2.0 mL, cryogenic
$630.00USD / Each
Corning FTS30 CoolCell Freezing System for Cryogenic Vials, green, 30 exposed vials; 1/ea EW-04392-08 Mfr # 432008
COMPARE
Green 30 1.0 to 2.0 mL, cryogenic
$630.00USD / Each
Corning FTS30 CoolCell Freezing System for Cryogenic Vials, pink, 30 exposed vials; 1/ea EW-04392-09 Mfr # 432009
COMPARE
Pink 30 1.0 to 2.0 mL, cryogenic
$630.00USD / Each
Showing 1- 10 of 10