Corning® CoolBox™ 2XT Ice-Free Coolers with Two Cooling Cores
No mess, no maintenance
  • Keep samples cold without melting ice contamination, inconvenient batteries, or power cord clutter
  • Cooling core maintains sample temperatures at 0.5°C to 4°C for at least four hours
  • Compatible with all Corning CoolSink® and CoolRack® sample modules (order separately)
These benchtop coolers eliminate possible contamination from ice, maintenance costs associated with battery replacement, and messy cords. Simply freeze the cooling core in a -20°C freezer for 12 or more hours, then remove and wait for core to reach 1°C before placing in holder. Once the core is installed, add your CoolSink or CoolRack module and load your samples. Samples quickly equilibrate to the core temperature and remain chilled for 4 to 16 hours.
Select the CoolBox system for longer-term cooling, which features a deeper sample area capable of maintaining temperature up to 16 hours.
$1,080.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color
Availability Pricing
Corning 2XT CoolBox™ Ice-Free Cooler with Two Cooling Cores, purple, 1/ea EW-04392-25 Mfr # 432025
COMPARE
Purple
$1,080.00USD / Each
Corning 2XT CoolBox™ Ice-Free Cooler with Two Cooling Cores, green, 1/ea EW-04392-26 Mfr # 432026
COMPARE
Green
$1,080.00USD / Each
Corning 2XT CoolBox™ Ice-Free Cooler with Two Cooling Cores, orange, 1/ea EW-04392-27 Mfr # 432027
COMPARE
Orange
$1,080.00USD / Each