This product is now discontinued, View Related Products

CoorsTek Pestles, Porcelain

Pestles are glazed all over except for grinding surfaces. Features an oversized pestle for comfort. Mortar and pestle are sold as separate pieces. Autoclavable.
Call For Price

14 variations of this product are available.

See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Pestles Length (mm)
Availability Pricing
CoorsTek 66311 Ceramic/Porcelain Pestle, 114 mm Length; 1/Pk EW-17821-00 Mfr # 66311
COMPARE
114
Call For Price
CoorsTek 66314 Ceramic/Porcelain Pestle, 130 mm Length; 1/Pk EW-17821-02 Mfr # 66314
COMPARE
130
Call For Price
CoorsTek 66317 Ceramic/Porcelain Pestle, 157 mm Length; 1/Pk EW-17821-04 Mfr # 66317
COMPARE
157
Call For Price
CoorsTek 66320 Ceramic/Porcelain Pestle, 180 mm Length; 1/Pk EW-17821-06 Mfr # 66320
COMPARE
180
Call For Price
CoorsTek 66323 Ceramic/Porcelain Pestle, 194 mm Length; 1/Pk EW-17821-08 Mfr # 66323
COMPARE
194
Call For Price
CoorsTek 66326 Ceramic/Porcelain Pestle, 222 mm Length; 1/Pk EW-17821-10 Mfr # 66326
COMPARE
222
Call For Price
CoorsTek 66329 Ceramic/Porcelain Pestle, 262 mm Length; 1/Pk EW-17821-12 Mfr # 66329
COMPARE
262
Call For Price
CoorsTek 60314 Pestle, Porcelain; 130 mm L, cs of 18 EW-17821-22 Mfr # 60314
COMPARE
130
Call For Price
CoorsTek 60317 Pestle, Porcelain; 157 mm L, cs of 12 EW-17821-24 Mfr # 66317
COMPARE
157
Call For Price
CoorsTek 60320 Pestle, Porcelain; 180 mm L, cs of 8 EW-17821-26 Mfr # 66320
COMPARE
180
Call For Price
CoorsTek 60323 Pestle, Porcelain; 194 mm L, cs of 8 EW-17821-28 Mfr # 66323
COMPARE
194
Call For Price
CoorsTek 60326 Pestle, Porcelain; 222 mm L, cs of 4 EW-17821-30 Mfr # 66326
COMPARE
222
Call For Price
CoorsTek 60329 Pestle, Porcelain; 262 mm L, cs of 2 EW-17821-32 Mfr # 66329
COMPARE
262
Call For Price
CoorsTek 66332 Ceramic/Porcelain Pestle, 280 mm Length; 1/Pk EW-17821-34 Mfr # 66332
COMPARE
280
Call For Price
Showing 1- 14 of 14

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat