This product is now discontinued, View Related Products

CoorsTek Pestles, Alumina

Ideal for precise elemental analysis where minimal sample contamination is required. Pestles are glazed all over except for grinding surfaces. Mortar and pestle are sold as separate pieces. Autoclavable.
Call For Price

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Pestles Length (mm)
Availability Pricing
CoorsTek 60356 Pestle, Alumina, 1.5 mL, 38 mm L EW-17823-00 Mfr # 60356
COMPARE
Call For Price
CoorsTek 60359 99.5% Alumina Pestle, 57 mm Length; 1/Pk EW-17823-02 Mfr # 60359
COMPARE
57
Call For Price
CoorsTek 60362 Pestle, Alumina, 15 mL, 65 31 mm L EW-17823-04 Mfr # 60362
COMPARE
Call For Price
CoorsTek 60359 Pestle, Alumina; 57 mm L, cs of 3 EW-17823-22 Mfr # 60359
COMPARE
57
Call For Price
CoorsTek 60365 Pestle, Alumina, 30 mL, 74 mm L EW-17823-26 Mfr # 60365
COMPARE
Call For Price
CoorsTek 60371 99.5% Alumina Pestle, 100 mm Length; 1/Pk EW-17823-28 Mfr # 60371
COMPARE
100
Call For Price
CoorsTek 60374 99.5% Alumina Pestle, 117 mm Length; 1/Pk EW-17823-30 Mfr # 60374
COMPARE
117
Call For Price
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat