Cooper-Atkins AquaTuff Waterproof Thermocouple Meters
Optimal choice for accuracy and maximum durability in food service applications
  • Large easy-to-read LCD
  • NSF, CE, RoHS, WEEE certified
The AquaTuff thermometer is a waterproof instrument built specifically for the harsh environments found in a commercial kitchen or food processing plant. The ABS housing is ergonomically designed with a large easy-to-read display and a curved back that fits nicely into your hand. The powerful microprocessor in this instrument delivers speed and reliability with a unique memory that stores the calibration settings, and reduces the need for frequent recalibration. Each instrument features a Fahrenheit/Celsius selection button, on/off button with auto-off after 10 minutes of non-use and easy twist-open water tight battery hatch.
The AquaTuff can be used with any Type-K thermocouple probe with a standard mini-connector, however food industry specific probes are listed below.
ITEM(S) ON SALE
$336.00 - $408.40USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Probe Channels Types Of Probes Accepted Description
Availability Pricing
Cooper-Atkins 35100-K Aquatuff Waterproof Thermocouple Meter EW-90025-07 Mfr # 35100-K
COMPARE
1 Type K Aquatuff Waterproof Thermocouple Meter
$336.00USD / Each
$420.00USD / Each
Cooper-Atkins 35200-K Aquatuff Advanced Waterproof Thermocouple Meter EW-90025-08 Mfr # 35200-K
COMPARE
1 Type K Aquatuff Advanced Waterproof Thermocouple Meter with Selectable Resolution and Hold Function
$408.40USD / Each
$510.50USD / Each