Comark Waterproof Thermocouple Thermometer
Ideal for use in food processing, HVAC, and industrial applications
  • IP67-rated case is water- and dustproof
  • Interactive menu allows for easy selection of all features
Thermometers are ideal for use in industrial, food processing, HVAC, refrigeration, and many more applications. They have a high degree of accuracy, rapid response time, and exceptional battery life. All models accept thermocouple probes with miniature connection, and feature an IP67-rated case, °F /°C selectable units, data hold, and auto-off function. Unit has BioCote® antimicrobial protection ideal for use in the food industry.
ITEM(S) ON SALE
$314.64USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
COMARK N9005 Waterproof Thermocouple Thermometer, Single Input EW-90025-25 Mfr # N9005
COMPARE
2501 1 Type K, Type T
$314.64USD / Each
$393.30USD / Each