Cole-Parmer® Essentials Watch Glasses
Manufactured from clear glass sheet (1.8 to 2.0mm thick), these watch glasses are fire ground and have polished edges. Concave shape prevents sliding and return of condensation to the beaker.
$15.50 - $287.00USD / Pkg of 12
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Fits Bottle / Beaker Size (mL) Outside Diameter (OD) (cm) Description
Availability Pricing
COMPARE
10 2.5 Watch Glass 25mm 12/pk
$15.50USD / Pkg of 12
COMPARE
30 4 Watch Glass 40mm 12/pk
$17.50USD / Pkg of 12
COMPARE
100 5 Watch Glass, 50 mm, fits 100 mL beakers, 12/pk
$19.50USD / Pkg of 12
COMPARE
150 6.5 Watch Glass, 65 mm, fits 150 mL beakers, 12/pk
$21.50USD / Pkg of 12
COMPARE
250 7.5 Watch Glass, 75 mm, fits 250 mL beakers, 12/pk
$22.50USD / Pkg of 12
COMPARE
400 9 Watch Glass, 90 mm, fits 400 mL beakers, 12/pk
$25.50USD / Pkg of 12
COMPARE
600 10 Watch Glass 100mm 12/pk
$36.80USD / Pkg of 12
COMPARE
1500 12.5 Watch Glass 125mm 12/pk
$42.00USD / Pkg of 12
COMPARE
3000 15 Watch Glass 150mm 12/pk
$57.30USD / Pkg of 12
COMPARE
4000 20 Watch Glass 200mm 12/pk
$66.50USD / Pkg of 12
COMPARE
7000 25 Watch Glass 250mm 12/pk
$185.00USD / Pkg of 12
COMPARE
11000 30 Watch Glass 300mm 12/pk
$287.00USD / Pkg of 12
Showing 1- 12 of 12