Cole-Parmer® VapLock™ Solvent Waste Manifold Caps for Panel Mounting
Connect solvent waste manifold caps to waste containers for a secure closed system
  • Choose from polypropylene or PTFE construction
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Panel-mount caps fit a 1-1/4" ID opening with panel thickness from 1/16" to 1/2" and comes with PP or PTFE locking nut. A 1-3/4" diameter obstruction-free area around the opening is required for use. Caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
$133.50 - $303.75USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material Special Features
Availability Pricing
COMPARE
Panel Mount Polypropylene(PP) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4\"-28
$167.50USD / Each
COMPARE
Panel Mount Polypropylene(PP) 1x1/2\" NPT, 3x1/4\" NPT
$133.50USD / Each
COMPARE
Panel Mount Polytetrafluoroethylene (PTFE) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4-28
$303.75USD / Each
COMPARE
Panel Mount Polytetrafluoroethylene (PTFE) 1x1/2\" NPT, 3x1/4\" NPT
$285.00USD / Each