Cole-Parmer® VapLock™ Solenoid-Operated 2- and 3-Way Pinch Valves
Open and close tubing to control fluid flow with minimal pressure drop
  • Quickly and easily change tubing between operation
  • No dead volume inside the valve
  • Unobstructed flow path—resists damage from particulates and easy to sterilize
  • Includes C-Flex® or silicone tubing
Solenoid-operated pinch valves open and close tubing to achieve controlled fluid flow. Since tubing is the only wetted part there is no dead volume and no obstruction in the flow path. Simply change the tubing when needed. Two-way valves are available in normally open or normally closed configurations. Three-way valves feature both a normally open and a normally closed flow path, which are operated simultaneously.
Designed for durability, valves last for a minimum 2 million open-and-close cycles and can remain in the fully energized state indefinitely without damage to the valve or tubing. PPS plunger and ASA/PC body material provide chemical resistance.
Valves are available with either C-Flex or platinum-cured silicone tubing. Both withstand the rigors of usage with pinch valves, will not stick even after prolonged closure, and are easily sterilizable. Tubing are rated for over 300,000 cycles. All two-way valves come with one 12"L tubing already in place. Three-way valves include two 6"L tubing joined by a Y connector.
†Low noise models 12021-25 and 12021-30 reduce the valve noise level from about 72 dB (A) to less than 50 dB (A).
‡Model 12021-40 features threaded mounting holes on the bottom of the valve body.
$103.00 - $217.00USD / Each
36 variations of this product are available.
See All Product Options Below
36 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Pinch Valve Tube Size Power (VDC)
Availability Pricing
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Closed, 1/32" ID C-Flex®; 12 VDC EW-12021-22 Mfr # 075P2NC12-01B
COMPARE
Two way, normally closed 1/32" ID x 3/32" OD 12
$114.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Closed, 1/32" ID Silicone; 12 VDC EW-12021-23 Mfr # 075P2NC12-01S
COMPARE
Two way, normally closed 1/32" ID x 3/32" OD 12
$122.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Closed, 1/16" ID Silicone; 12 VDC EW-12021-24 Mfr # 075P2NC12-02S
COMPARE
Two way, normally closed 1/16" ID x 1/8" OD 12
$114.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Closed, 1/32" ID Silicone; 24 VDC EW-12021-28 Mfr # 075P2NC24-01S
COMPARE
Two way, normally closed 1/32" ID x 3/32" OD 24
$123.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Closed, 1/16" ID Silicone; 24 VDC EW-12021-29 Mfr # 075P2NC24-02S
COMPARE
Two way, normally closed 1/16" ID x 1/8" OD 24
$114.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Closed, 1/16" ID Silicone, Silenced; 24 VDC EW-12021-30 Mfr # 075P2NC24-02SQ
COMPARE
Two way, normally closed 1/16" ID x 1/8" OD 24
$122.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Open, 1/32" ID Silicone; 12 VDC EW-12021-33 Mfr # 075P2NO12-01S
COMPARE
Two way, normally open 1/32" ID x 3/32" OD 12
$124.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Open, 1/32" ID Silicone; 24 VDC EW-12021-36 Mfr # 075P2NO24-01S
COMPARE
Two way, normally open 1/32" ID x 3/32" OD 24
$124.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Two-Way Normally Open, 1/16" ID Silicone; 24 VDC EW-12021-37 Mfr # 075P2NO24-02S
COMPARE
Two way, normally open 1/16" ID x 1/8" OD 24
$124.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Three-Way, 1/32" ID C-Flex®; 12 VDC EW-12021-38 Mfr # 075P3MP12-01B
COMPARE
Three way, one port normally open, other normally closed 1/32" ID x 3/32" 12
$151.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Three-Way, 1/32" ID Silicone, Mounting Holes; 12 VDC EW-12021-40 Mfr # 075P3MP12-01SM
COMPARE
Three way, one port normally open, other normally closed 1/32" ID x 3/32" OD 12
$186.00USD / Each
Cole-Parmer VapLock™ Solenoid Operated Pinch Valve, Three-Way, 1/32" ID Silicone; 24 VDC EW-12021-43 Mfr # 075P3MP24-01S
COMPARE
Three way, one port normally open, other normally closed 1/32" ID x 3/32" OD 24
$151.00USD / Each
Cole-Parmer VapLock™ 2-Way Pinch Valve, Normally Closed, Silicone Tubing, 1/16" Inner Diameter, 1/8" Outer Diameter, 12 VDC EW-12021-68 Mfr # 100P2NC12-02S
COMPARE
2-way, normally closed 1/16" ID x 1/8" OD 12
$159.00USD / Each
Cole-Parmer VapLock™ 2-Way Pinch Valve, Normally Closed, Silicone Tubing, 1/8" Inner Diameter, 1/4" Outer Diameter, 12 VDC EW-12021-71 Mfr # 100P2NC12-05S
COMPARE
2-way, normally closed 1/8" ID x 1/4" OD 12
$171.00USD / Each
Cole-Parmer VapLock™ 2-Way Pinch Valve, Normally Closed, C-Flex® Tubing, 1/8" Inner Diameter, 1/4" Outer Diameter, 24 VDC EW-12021-75 Mfr # 100P2NC24-05B
COMPARE
2-way, normally closed 1/8" ID x 1/4" OD 24
$159.00USD / Each
Cole-Parmer VapLock™ 2-Way Pinch Valve, Normally Closed, Silicone Tubing, 1/8" Inner Diameter, 1/4" Outer Diameter, 24 VDC EW-12021-76 Mfr # 100P2NC24-05S
COMPARE
2-way, normally closed 1/8" ID x 1/4" OD 24
$103.00USD / Each
Cole-Parmer VapLock™ 2-Way Pinch Valve, Normally Open, Silicone Tubing, 1/8" Inner Diameter, 1/4" Outer Diameter, 12 VDC EW-12021-78 Mfr # 100P2NO12-05S
COMPARE
2-way, normally open 1/8" ID x 1/4" OD 12
$168.00USD / Each
Cole-Parmer VapLock™ 3-Way Pinch Valve, One Port Normally Open, Other Port Normally Closed, Silicone Tubing, 1/16" Inner Diameter, 1/8" Outer Diameter, 12 VDC EW-12021-80 Mfr # 100P3MP12-02S
COMPARE
3-way 1/16" ID x 1/8" OD 12
$187.00USD / Each
Cole-Parmer VapLock™ 3-Way Pinch Valve, One Port Normally Open, Other Port Normally Closed, Silicone Tubing, 1/8" Inner Diameter, 1/4" Outer Diameter, 12 VDC EW-12021-83 Mfr # 100P3MP12-05S
COMPARE
3-way 1/8" ID x 1/4" OD 12
$217.00USD / Each
Cole-Parmer VapLock™ 3-Way Pinch Valve, One Port Normally Open, Other Port Normally Closed, Silicone Tubing, 1/8" Inner Diameter, 1/4" Outer Diameter, 24 VDC EW-12021-85 Mfr # 100P3MP24-05S
COMPARE
3-way 1/8" ID x 1/4" OD 24
$217.00USD / Each
Showing 1 - 20 of 36