Cole-Parmer OS-200 Series Ultra-Compact Digital Mixer System
Complete digital system for your general mixing needs—includes mixer, stand, and two blades
  • Compact size—only 2”W x 4”H x 6”D—fits virtually anywhere!
  • Mixes up to 2 liters of water at a maximum speed of 2500 rpm
This compact, digital mixer is perfect for smaller scale mixing needs throughout your laboratory. The maintenance-free brushless DC motor provides speeds from 50 to 2500 rpm. The LED display lets you make precise speed adjustments and ensures repeatability. The motor housing is lightweight molded nylon, resistant to the occasional splash and spill.
Mixer housing includes an integrated stand clamp that allows for quick and secure mounting to the included stand. The stainless steel collet, with set screws, accepts 1/4" (6-mm) shafts.
$552.00 - $558.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
COMPARE
2500 14.2 115
$552.00USD / Each
COMPARE
2500 14.2 100 to 240
$558.00USD / Each