Cole-Parmer Twistit Stoppers, Rubber
Use these versatile Twistit stoppers as either a solid, one-hole, two-hole, or three-hole stopper. Simply twist off a bottom seal to open a hole. The soft resilient natural rubber stays pliable over a long period and withstands laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Available in black or white rubber. Autoclavable.
$61.30 - $74.00USD / Pkg of 35
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Standard Size Number Description
Availability Pricing
COMPARE
Rubber 3 Twistit black rubber stopper, size 3
$61.30USD / Pkg of 50
COMPARE
Rubber 4 Twistit black rubber stopper, size 4
$61.30USD / Pkg of 40
COMPARE
Rubber 5 Twistit black rubber stopper, size 5
$61.30USD / Pkg of 35
COMPARE
Rubber 6 Twistit black rubber stopper, size 6
$61.30USD / Pkg of 25
COMPARE
Rubber 7 Twistit black rubber stopper, size 7
$61.30USD / Pkg of 20
COMPARE
Rubber 8 Twistit black rubber stopper, size 8
$61.30USD / Pkg of 16
COMPARE
Rubber 9 Twistit black rubber stopper, size 9
$61.30USD / Pkg of 12
COMPARE
Rubber 10 Twistit black rubber stopper, size 10
$61.30USD / Pkg of 10
COMPARE
Rubber Twistit black rubber stopper assortment pack
$62.60USD / Pkg of 33
COMPARE
Rubber 3 Twistit white rubber stopper, size 3
$64.60USD / Pkg of 50
COMPARE
Rubber 4 Twistit white rubber stopper, size 4
$74.00USD / Pkg of 40
COMPARE
Rubber 6 Twistit white rubber stopper, size 6
$73.50USD / Pkg of 25
COMPARE
Rubber 7 Twistit white rubber stopper, size 7
$72.80USD / Pkg of 20
COMPARE
Rubber 8 Twistit white rubber stopper, size 8
$72.10USD / Pkg of 16
COMPARE
Rubber 9 Twistit white rubber stopper, size 9
$71.40USD / Pkg of 12
COMPARE
Rubber 10 Twistit white rubber stopper, size 10
$70.80USD / Pkg of 10
COMPARE
Rubber Twistit white rubber stopper assortment pack
$71.40USD / Pkg of 33
Showing 1- 17 of 17