This product is now discontinued, View Related Products
Cole-Parmer Tube Roller Shakers
Get complete mixing with both rocking and rolling movement
  • Use in cold rooms down to 39°F (4°C) or in incubators up to 140°F (60°C) with 80% humidity
  • Gentle, continuous, and quiet operation
These solidly constructed tube roller shakers are ideal for mixing samples where minimum aeration is required — like viscous substances or liquid-solid suspensions. Save bench space while expanding your capacity with the stacking kit (sold separately), and stack up to three shakers!
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
33 230
Call For Price
COMPARE
33 120
Call For Price
COMPARE
60 230
Call For Price
COMPARE
60 120
Call For Price
COMPARE
33 230
Call For Price
COMPARE
33 120
Call For Price
COMPARE
60 230
Call For Price
COMPARE
60 120
Call For Price
Showing 1- 8 of 8