This product is now discontinued, View Related Products
Cole-Parmer Tube Rockers
Achieve uniform mixing while preserving sample integrity
  • Pause button halts rocking motion while adding or removing tubes, extending the life of the motor
  • 25 rocking cycles per minute provide gentle mixing
Heavy-duty tube rockers use minimal benchtop space and provide years of everyday use. The uniform 30° rocking angle provides smooth and gentle motion for a thoroughly mixed sample. Reversible non-slip pad will accommodate tubes up to 120-mm long and is autoclavable.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Number of Tubes Power (VAC)
Availability Pricing
COMPARE
8 110
Call For Price
COMPARE
8 220
Call For Price
COMPARE
15 110
Call For Price
COMPARE
15 220
Call For Price
COMPARE
30 110
Call For Price
COMPARE
30 220