Cole-Parmer Toroidal Conductivity Transmitters
Cole-Parmer® Toroidal Conductivity Transmitters
  • In-line or submersible installation
  • CPVC and PVDF models available
  • Built in 100 Ω Pt RTD temperature compensation
The electrodeless conductivity sensor is coupled with a two-wire, loop-powered 4 to 20 mA transmitter. The sensor requires an 11 to 24 VDC power supply and has a maximum draw of 600 ohms. If 11 to 24 VDC power is not already available, order process meter 94785-12 to power and display the sensor measurements, or refer to our process meter section for other models.
The isolated 4 to 20 mA output can be used for control applications, connected to a recorder, or an alarm. The sensor has a temperature coefficient of 0.3%/degC, and measures 8''L x 1-1/2'' diameter with 4-1/2'' insertion depth. Sensor comes with a 10-ft cable with tinned ends.
$595.00 - $862.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Toroidal Conductivity transmitter, CPVC, 0 to 10 mS
$599.00USD / Each
COMPARE
Toroidal Conductivity transmitter, CPVC, 0 to 100 mS
$595.00USD / Each
COMPARE
Toroidal Conductivity transmitter, CPVC, 0 to 1000 mS
$595.00USD / Each
COMPARE
Toroidal Conductivity transmitter, PVDF, 0 to 100 mS
$862.00USD / Each
COMPARE
Toroidal Conductivity transmitter, PVDF, 0 to 1000 mS
$831.50USD / Each