This product is now discontinued, View Related Products
Cole-Parmer® Symmetry™ ISW Washdown Industrial Bench Scales
IP66 rated for washdown use in agricultural, industrial, and food processing facilities
  • Color-coded keys provide quick access to weighing units (kg, g, oz, lb, and lb:oz)
  • Programmable backlit LCD with capacity tracker easily monitors possible overloads
  • Overload protection helps prevent internal damage
  • Auto power-off function reduces power consumption
  • Rugged 304 stainless steel and ABS construction withstand harsh environments
  • Functions include weighing and checkweighing
These lightweight ISW washdown bench scales feature an IP66-rated housing with a large, easy-to-clean stainless steel weighing pan. Checkweighing is quick and easy with built-in color indicators that tell you if the load is above, below, or within preset limits. The scales include automatic zero tracking, audible alarm for pre-set weights, low battery indicator, semi-automatic tare, and adjustable leveling feet.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
COMPARE
3000 115 0.5
Call For Price
COMPARE
6000 115 1
Call For Price
COMPARE
12000 115 2
Call For Price
COMPARE
3000 220 0.5
Call For Price
COMPARE
6000 220 1
Call For Price
COMPARE
12000 220 2