Cole-Parmer® ST-300 Series Digital Round-Top Stirrers
Simple and precise stirring at your fingertips
  • Innovative fingertip control even while wearing gloves
  • Advanced brushless motor provides quiet and worry-free operation
  • Sealed exterior keeps spills away for easy maintenance
  • LCD shows set and working parameters
  • Ceramic-coated aluminum surface offers a robust top plate
Sleek, low-profile digital stirrer has a bright LCD showing both set and working parameters which are lockable to prevent accidental changes during use. Unique control pad allows all operations to be set using just your fingertip.
Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built-in timer can be set for up to 9 hours or continuous operation. Unit offers stirring speeds up to 1800 rpm with max capacity of 20 liters.
The ceramic-coated aluminum top plate offers excellent chemical resistance and is easy to clean.
$430.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Power (VAC) Top Plate Diameter (in)
Availability Pricing
COMPARE
1800 110 5 19/64
$430.00USD / Each
COMPARE
1800 220 5 19/64
$430.00USD / Each