Cole-Parmer StableTemp Dry Block Heaters (Digital)

Precision temperature stability ensures reproducibility of results
  • Create a versatile system unique to your application; order a digital heater and the required heating block module (sold separately online).
  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes -order blocks separately
Designed for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures. Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes. Modular blocks are constructed from a solid anodized aluminum block. The close tube to block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Optional PT100 RTD temperature probe kit enables the unit to read the actual block or sample temperature and display the temperature on the control panel. Create a versatile system unique to your application; order either an analog or digital heater and the required heating block module (sold separately online).

CAUTION! To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed to run as a dry bath / incubator.

Digital Dry Block Heaters offer exceptional temperature uniformity and feature a proportional integrative derivative (PID) controller. PID control allows you to select a particular temperature with ease (in 0.1°C increments), enables rapid heat-up for samples, and provides excellent stability to maintain results. Features include timer, audible alarm, built-in temperature calibration mode, overshoot protection, and independent LED displays that show actual and setpoint temperatures
$687.00 - $796.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Number Of Blocks Power (VAC)
Availability Pricing
COMPARE
12 2/5 1 120
$687.00USD / Each
COMPARE
12 2/5 1 230
$687.00USD / Each
COMPARE
15 2/5 2 120
$796.00USD / Each
COMPARE
15 2/5 2 230
$796.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat