Cole-Parmer Specific Gravity Combined Form Precision Hydrometers for Liquids Heavier than Water
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
ITEM(S) ON SALE
$53.29 - $58.56USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
1.000/1.220 Specific Gravity Combined Form Thermo-Hydrometer
$53.29USD / Each
$62.70USD / Each
COMPARE
1.200/1.420 Specific Gravity Combined Form Thermo-Hydrometer
$54.14USD / Each
$63.70USD / Each
COMPARE
1.400/1.620 Specific Gravity Combined Form Thermo-Hydrometer
$54.99USD / Each
$64.70USD / Each
COMPARE
1.600/1.820 Specific Gravity Combined Form Thermo-Hydrometer
$57.20USD / Each
$67.30USD / Each
COMPARE
1.800/2.000 Specific Gravity Combined Form Thermo-Hydrometer
$58.56USD / Each
$68.90USD / Each