Cole-Parmer Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Conform to ASTM Specifications
  • Metal Ballast
  • Individually Serialized
These hydrometers conform to the accuracy requirements for ASTM hydrometers. Scales are affixed with clear epoxy. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
ITEM(S) ON SALE
$43.01 - $66.47USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0.650/0.700 Specific Gravity ASTM 82H Hydrometer for Light Liquids
$43.01USD / Each
$50.60USD / Each
COMPARE
0.700/0.750 Specific Gravity ASTM 83H Hydrometer for Light Liquids
$48.02USD / Each
$56.50USD / Each
COMPARE
0.750/0.800 Specific Gravity ASTM 84H Hydrometer for Light Liquids
$44.88USD / Each
$52.80USD / Each
COMPARE
0.800/0.850 Specific Gravity ASTM 85H Hydrometer for Light Liquids
$48.45USD / Each
$57.00USD / Each
COMPARE
0.850/0.900 Specific Gravity ASTM 86H Hydrometer for Light Liquids
$47.60USD / Each
$56.00USD / Each
COMPARE
0.900/0.950 Specific Gravity ASTM 87H Hydrometer for Light Liquids
$48.45USD / Each
$57.00USD / Each
COMPARE
0.950/1.000 Specific Gravity ASTM 88H Hydrometer for Light Liquids
$47.60USD / Each
$56.00USD / Each
COMPARE
1.000/1.050 Specific Gravity ASTM 89H Hydrometer for Light Liquids
$48.45USD / Each
$57.00USD / Each
COMPARE
1.050/1.100 Specific Gravity ASTM 90H Hydrometer for Light Liquids
$66.47USD / Each
$78.20USD / Each
Showing 1- 9 of 9