Cole-Parmer Sealed Stainless Steel Vacuum and Compound Transducers
Superior long-term stability and repeatability
  • Feature 4 to 20 mA output and have red (positive power) and black (output) wires
Stainless steel diaphragm and polysilicon strain gauge are bonded by vapor deposition—no epoxies or glues which can cause signal instability or drift.
ITEM(S) ON SALE
$348.00 - $435.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±0.4% (BFSL)
$348.00USD / Each
$435.00USD / Each
COMPARE
±0.4% (BFSL) 30
$348.00USD / Each
$435.00USD / Each
COMPARE
±0.4% (BFSL) 60
$348.00USD / Each
$435.00USD / Each
COMPARE
±0.4% (BFSL)
$435.00USD / Each
COMPARE
±0.4% (BFSL) 30
$435.00USD / Each