Fisher Chemical™ Potassium Chloride

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$174.00 - $950.50USD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Weight (g) Description
Availability Pricing
Fisher Chemical Potassium Chloride, Certified ACS Grade, 500 g EW-88019-84 Mfr # P217500
COMPARE
Certified ACS 500 Potassium Chloride, Certified ACS Grade, 500 g
$212.75USD / Each
Fisher Chemical Potassium Chloride, Certified ACS Grade, 3 kg EW-88019-86 Mfr # P2173
COMPARE
Certified ACS 3000 Potassium Chloride, Certified ACS Grade, 3 kg
$950.50USD / Each
Fisher Chemical Potassium Chloride, BP/EP/FCC/JP/USP Grade, 2.5 kg EW-88019-88 Mfr # P335212
COMPARE
BP/EP/FCC/JP/USP 2500 Potassium Chloride, BP/EP/FCC/JP/USP Grade, 2.5 kg
$736.50USD / Each
Fisher Chemical Potassium Chloride (White Crystals) (1kg) EW-88066-92 Mfr # BP3661
COMPARE
None 1000 Potassium Chloride (White Crystals) (1kg)
$413.75USD / Each
Fisher Chemical Potassium Chloride (White Crystals) (500g) EW-88066-93 Mfr # BP366500
COMPARE
None 500 Potassium Chloride (White Crystals) (500g)
$219.75USD / Each
Fisher Chemical Potassium Chloride, FCC/USP Grade, 3 kg EW-88079-05 Mfr # P3303
COMPARE
FCC/USP 3000 Potassium Chloride, FCC/USP Grade, 3 kg
$563.00USD / Each
Fisher Chemical Potassium Chloride, FCC/USP Grade, 500 g EW-88079-06 Mfr # P330500
COMPARE
FCC/USP 500 Potassium Chloride, FCC/USP Grade, 500 g
$174.00USD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat