Cole-Parmers® IP-400 Series Low-Temperature Immersion Probe Chillers
Quickly chill liquids in container or open baths
  • Economical alternative to dry ice or liquid nitrogen
  • Ideal for freeze drying, rapid fluid cooling, and trapping applications
  • Designed to run at maximum cooling
Ideal for use with circulating baths, these chillers provide rapid, low-cost cooling of liquids to temperatures as low as -100°C. Applications include cannabis extraction, cooling of exothermic reactions, freeze point determinations, botanical extraction, freeze drying, impact testing, lyophilization, and vapor and solvent trapping. The chillers are a cost-effective alternative to dry ice or liquid nitrogen.
Simple to use. Just plug it in and immerse the stainless-steel probe in your fluid sample. A flexible hose allows convenient placement of the cooling probe. Please note, these chillers do not have an adjustable low temperature set point.
$2,509.00 - $7,707.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Power (VAC) Wetted Materials
Availability Pricing
COMPARE
-60 120 Rigid Coil Probe
$2,509.00USD / Each
COMPARE
-60 240 Rigid Coil Probe
$2,509.00USD / Each
COMPARE
-35 120 Rigid Coil Probe
$3,299.00USD / Each
COMPARE
-35 240 Rigid Coil Probe
$3,299.00USD / Each
COMPARE
-100 120 Flexible Cold Finger Probe
$7,125.00USD / Each
COMPARE
-100 120 Rigid Coil Probe
$7,125.00USD / Each
COMPARE
-100 240 Rigid Coil Probe
$7,690.00USD / Each
COMPARE
-80 120 Rigid Coil Probe
$7,707.00USD / Each
COMPARE
-80 240 Rigid Coil Probe
$7,707.00USD / Each
Showing 1- 9 of 9